Anti-TLK1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58340_P050
AVIVARP58340_P050EA 401 EUR
AVIVARP58340_P050
Anti-TLK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TLK1 (tousled-like kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen