Anti-CDK1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58401_P050
AVIVARP58401_P050EA 401 EUR
AVIVARP58401_P050
Anti-CDK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human