Anti-ALDH2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58418_P050
AVIVARP58418_P050EA 401 EUR
AVIVARP58418_P050
Anti-ALDH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen