Anti-CAT Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58437_P050
AVIVARP58437_P050EA 401 EUR
AVIVARP58437_P050
Anti-CAT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAT (catalase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen