Anti-GAPDH Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58579_P050
AVIVARP58579_P050EA 401 EUR
AVIVARP58579_P050
Anti-GAPDH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen