Anti-NOG Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP58773_P050
AVIVARP58773_P050EA 401 EUR
AVIVARP58773_P050
Anti-NOG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOG (noggin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen