Anti-SPATA2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59151_P050
AVIVARP59151_P050EA 401 EUR
AVIVARP59151_P050
Anti-SPATA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPATA2 (spermatogenesis associated 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen