Anti-SERPINA10 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP59235_P050
AVIVARP59235_P050EA 401 EUR
AVIVARP59235_P050
Anti-SERPINA10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA10 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR