Anti-CDC16 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59260_P050
AVIVARP59260_P050EA 401 EUR
AVIVARP59260_P050
Anti-CDC16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC16 (cell division cycle 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen