Anti-SNPH Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59516_P050
AVIVARP59516_P050EA 401 EUR
AVIVARP59516_P050
Anti-SNPH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNPH (syntaphilin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen