Anti-ELMO1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59647_P050
AVIVARP59647_P050EA 401 EUR
AVIVARP59647_P050
Anti-ELMO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen