Anti-OR2L8 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59738_P050
AVIVARP59738_P050EA 401 EUR
AVIVARP59738_P050
Anti-OR2L8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR2L8 (olfactory receptor, family 2, subfamily L, member 8 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human