Anti-CANX Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59755_P050
AVIVARP59755_P050EA 401 EUR
AVIVARP59755_P050
Anti-CANX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CANX (calnexin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen