Anti-ALDH1A2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP59783_P050
AVIVARP59783_P050EA 401 EUR
AVIVARP59783_P050
Anti-ALDH1A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH1A2 (aldehyde dehydrogenase 1 family, member A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen