Anti-ENOX2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP59871_P050
AVIVARP59871_P050EA 401 EUR
AVIVARP59871_P050
Anti-ENOX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ENOX2 (ecto-NOX disulfide-thiol exchanger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR