Anti-CLTA Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP59988_P050
AVIVARP59988_P050EA 401 EUR
AVIVARP59988_P050
Anti-CLTA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLTA (clathrin, light chain A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR