Anti-OVGP1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP60216_P050
AVIVARP60216_P050EA 401 EUR
AVIVARP60216_P050
Anti-OVGP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OVGP1 (oviductal glycoprotein 1, 120kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human