Anti-BCAM Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP60229_P050
AVIVARP60229_P050EA 401 EUR
AVIVARP60229_P050
Anti-BCAM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCAM (basal cell adhesion molecule (Lutheran blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen