Anti-MARCKS Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP60325_P050
AVIVARP60325_P050EA 401 EUR
AVIVARP60325_P050
Anti-MARCKS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human