Anti-CAPN3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP60947_P050
AVIVARP60947_P050EA 401 EUR
AVIVARP60947_P050
Anti-CAPN3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAPN3 (calpain 3, (p94))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen