Anti-H2AFZ Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP61353_P050
AVIVARP61353_P050EA 401 EUR
AVIVARP61353_P050
Anti-H2AFZ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFZ (H2A histone family, member Z)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen