Anti-AKR1C3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP61430_P050
AVIVARP61430_P050EA 401 EUR
AVIVARP61430_P050
Anti-AKR1C3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen