Anti-SPC25 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP61828_P050
AVIVARP61828_P050EA 401 EUR
AVIVARP61828_P050
Anti-SPC25 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPC25 (SPC25, NDC80 kinetochore complex component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol SPC25
Antigen-Name SPC25, NDC80 kinetochore complex component
Environmentally Preferable
Antigen-Synonyme SPBC25|AD024|hSpc25
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Pig,Human,Bovine,Dog,Rat,Mouse
Wirt Rabbit
Gen-ID 57405
Isotyp
Western blot Yes
Epitop middle
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR