Anti-NUP160 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP61864_P050
AVIVARP61864_P050EA 401 EUR
AVIVARP61864_P050
Anti-NUP160 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUP160 (nucleoporin 160kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen