Anti-PFKFB1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP61878_P050
AVIVARP61878_P050EA 401 EUR
AVIVARP61878_P050
Anti-PFKFB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PFKFB1 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen