Anti-MED18 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP62133_P050
AVIVARP62133_P050EA 401 EUR
AVIVARP62133_P050
Anti-MED18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MED18 (mediator complex subunit 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR