Anti-TPD52L2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62163_P050
AVIVARP62163_P050EA 401 EUR
AVIVARP62163_P050
Anti-TPD52L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPD52L2 (tumor protein D52-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen