Anti-CTPS2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62443_P050
AVIVARP62443_P050EA 401 EUR
AVIVARP62443_P050
Anti-CTPS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTPS2 (CTP synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen