Anti-MRPS11 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62568_P050
AVIVARP62568_P050EA 401 EUR
AVIVARP62568_P050
Anti-MRPS11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MRPS11 (mitochondrial ribosomal protein S11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen