Anti-SFXN2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62590_P050
AVIVARP62590_P050EA 401 EUR
AVIVARP62590_P050
Anti-SFXN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SFXN2 (sideroflexin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen