Anti-GPR52 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP62835_P050
AVIVARP62835_P050EA 401 EUR
AVIVARP62835_P050
Anti-GPR52 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPR52 (G protein-coupled receptor 52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR