Anti-LYZL4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62861_P050
AVIVARP62861_P050EA 401 EUR
AVIVARP62861_P050
Anti-LYZL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYZL4 (lysozyme-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat