Anti-TARS2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP62923_P050
AVIVARP62923_P050EA 401 EUR
AVIVARP62923_P050
Anti-TARS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TARS2 (threonyl-tRNA synthetase 2, mitochondrial (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen