Anti-CFI Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP63227_P050
AVIVARP63227_P050EA 401 EUR
AVIVARP63227_P050
Anti-CFI Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CFI (complement factor I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human