Anti-TPMT Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP63262_P050
AVIVARP63262_P050EA 401 EUR
AVIVARP63262_P050
Anti-TPMT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPMT (thiopurine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Pig
Bestellen

TECHNISCHE DATEN

Antigen-Symbol TPMT
Environmentally Preferable
Antigen-Name Thiopurine S-methyltransferase
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Pig,Human
Wirt Rabbit
Gen-ID 7172
Isotyp
Western blot Yes
Epitop C-terminal
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR