Anti-KLRK1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP63274_P050
AVIVARP63274_P050EA 401 EUR
AVIVARP63274_P050
Anti-KLRK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLRK1 (killer cell lectin-like receptor subfamily K, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR