Anti-KLRD1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP63304_P050
AVIVARP63304_P050EA 401 EUR
AVIVARP63304_P050
Anti-KLRD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLRD1 (killer cell lectin-like receptor subfamily D, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human