Anti-EXO1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP63489_P050
AVIVARP63489_P050EA 401 EUR
AVIVARP63489_P050
Anti-EXO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXO1 (exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen