Anti-CD151 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP63500_P050
AVIVARP63500_P050EA 401 EUR
AVIVARP63500_P050
Anti-CD151 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD151 (CD151 molecule (Raph blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR