Anti-ICK Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP63766_P050
AVIVARP63766_P050EA 401 EUR
AVIVARP63766_P050
Anti-ICK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ICK (intestinal cell (MAK-like) kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human