Anti-STRA8 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP63959_P050
AVIVARP63959_P050EA 401 EUR
AVIVARP63959_P050
Anti-STRA8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STRA8 (stimulated by retinoic acid 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen