Anti-DKFZp779C2259 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP63988_P050
AVIVARP63988_P050EA 401 EUR
AVIVARP63988_P050
Anti-DKFZp779C2259 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DKFZp779C2259
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR