Anti-ARL1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP64038_P050
AVIVARP64038_P050EA 401 EUR
AVIVARP64038_P050
Anti-ARL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARL1 (ADP-ribosylation factor-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen