Anti-DUSP4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP64084_P050
AVIVARP64084_P050EA 401 EUR
AVIVARP64084_P050
Anti-DUSP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DUSP4 (dual specificity phosphatase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen