Anti-RPS17 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP64464_P050
AVIVARP64464_P050EA 401 EUR
AVIVARP64464_P050
Anti-RPS17 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS17 (ribosomal protein S17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human