Anti-NAPSA Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP64545_P050
AVIVARP64545_P050EA 401 EUR
AVIVARP64545_P050
Anti-NAPSA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAPSA (napsin A aspartic peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR