Anti-NPR3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP64738_P050
AVIVARP64738_P050EA 401 EUR
AVIVARP64738_P050
Anti-NPR3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPR3 (natriuretic peptide receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR