Anti-RPS11 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65143_P050
AVIVARP65143_P050EA 401 EUR
AVIVARP65143_P050
Anti-RPS11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS11 (ribosomal protein S11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human