Anti-CAPRIN1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP65220_P050
AVIVARP65220_P050EA 401 EUR
AVIVARP65220_P050
Anti-CAPRIN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAPRIN1 (cell cycle associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR