Anti-CLNK Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP65309_P050
AVIVARP65309_P050EA 401 EUR
AVIVARP65309_P050
Anti-CLNK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLNK (cytokine-dependent hematopoietic cell linker)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human